Methyl-3-oxoheptanoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Oxoheptanoate, 3-Ketoheptanoic Acid Methyl Ester, 3-Oxoheptanoic Acid Methyl Ester, 3-Oxoenanthic Acid Methyl Ester, Methyl 3-Oxoenanthate, Methyl 3-oxoheptanoate

A150604-100G A150604-10G A150604-25G A150604-5G
103804-186EA 9 USD
103804-186 103842-322 76719-708 76719-710 77299-816 77317-850
Methyl-3-oxoheptanoate 95%
Methyl-3-oxoheptanoate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.20 g/mol
Boiling Pt: 206 °C (760 mm Hg)
Flash Pt: 79.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00191568
CAS Number: 39815-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR