Methyl 3-oxocyclohex-1-enecarboxylate 95%

Supplier: Ambeed
Warning

A162430-100MG A162430-250MG A162430-5G A162430-1G
103805-614EA 135 USD
103805-614 76720-154 76720-156 76720-158 77319-908
Methyl 3-oxocyclohex-1-enecarboxylate 95%
Methyl 3-oxocyclohex-1-enecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11053675
CAS Number: 54396-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR