Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-oxo-4-phenylbutanoate, Methyl 4-Phenylacetoacetate, 3-Oxo-4-phenylbutyric Acid Methyl Ester, 4-Phenylacetoacetic Acid Methyl Ester

A183799-1G A183799-25G A183799-5G A183799-10G
103815-670EA 32 USD
103815-670 103846-232 77301-858 77301-860 77301-862 77667-528
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers) 95%
Methyl-3-oxo-4-phenylbutyrate (mixture of isomers)

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 273.4 °C (760 mm Hg)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD03424746
CAS Number: 37779-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR