Methyl 3-oxo-4-(2,4,5-trifluorophenyl)butanoate 97%

Supplier: Ambeed
Warning

A137850-100G A137850-10G A137850-250MG A137850-25G A137850-5G A137850-1G
103802-516EA 7 USD
103802-516 76972-426 76972-428 76972-430 76972-432 77298-928 77315-498
Methyl 3-oxo-4-(2,4,5-trifluorophenyl)butanoate 97%
Methyl 3-oxo-4-(2,4,5-trifluorophenyl)butanoate

Precaución: for Research Use Only

Formula: C₁₁H₉F₃O₃
MW: 246.183 g/mol
Boiling Pt: 273.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09833197
CAS Number: 769195-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR