Methyl-3-methylthiophene-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-methylthiophene-2-carboxylate
A150394-100G
A150394-10G
A150394-1G
A150394-25G
103804-156EA
12
USD
103804-156
103842-290
76768-138
76768-140
77299-796
77299-798
Methyl-3-methylthiophene-2-carboxylate 98%
Methyl 3-methylthiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₈O₂S Boiling Pt: 211 °C (760 mm Hg) Flash Pt: 81.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234332 CAS Number: 81452-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...