Methyl-3-methylpicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Methylpicolinate, Methyl 3-Methylpyridine-2-carboxylate, Methyl 3-methylpicolinate, 3-Methylpyridine-2-carboxylic acid methyll ester, 3-Methylpicolinic acid methyll ester

A586467-100G A586467-10G A586467-5G A586467-1G A586467-25G
103828-892EA 6 USD
103828-892 76726-768 76726-770 76726-772 77307-880 77594-776
Methyl-3-methylpicolinate 98%
Methyl 3-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Boiling Pt: 241.1 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03789583
CAS Number: 59718-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR