Methyl 3-methyloxetane-3-carboxylate 97%

Supplier: Ambeed
Warning

A236807-250MG A236807-5G A236807-1G
103811-118EA 342 USD
103811-118 76987-524 76987-526 77304-062
Methyl 3-methyloxetane-3-carboxylate 97%
Methyl 3-methyloxetane-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.142 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18251059
CAS Number: 1260670-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR