Methyl-3-methyl-5-nitrobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-methyl-5-nitrobenzoate
A152953-250MG
A152953-1G
103804-454EA
92
USD
103804-454
103842-626
76984-702
77318-260
Methyl-3-methyl-5-nitrobenzoate 97%
Methyl 3-methyl-5-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₉H₉NO₄ MW: 195.1700 g/mol Boiling Pt: 315.0±22.0 °C (760 mm Hg) Melting Pt: 84…85 °C Flash Pt: 148.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD12172988 CAS Number: 482311-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...