Methyl 3-methyl-1H-pyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

A620749-250MG A620749-25G A620749-1G A620749-100MG
103840-214EA 237 USD
103840-214 76980-620 76980-622 77307-968 77673-680
Methyl 3-methyl-1H-pyrrole-2-carboxylate 97%
Methyl 3-methyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂
Boiling Pt: 232.0±20.0 °C (760 mm Hg)
Flash Pt: 94.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09754196
CAS Number: 40611-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR