Methyl-3-methyl-1H-pyrazole-4-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-methyl-1H-pyrazole-4-carboxylate
A113419-250MG
A113419-1G
103798-228EA
33
USD
103798-228
103824-070
76971-010
77297-244
Methyl-3-methyl-1H-pyrazole-4-carboxylate 95%
Methyl-3-methyl-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ MW: 140.1400 g/mol Boiling Pt: 268.5 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD07357354 CAS Number: 23170-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...