Methyl m-anisate 97%

Supplier: Ambeed
Warning

A238554-100G A238554-10G A238554-1G A238554-500G A238554-25G A238554-5G
103851-018EA 6 USD
103851-018 76722-384 76722-386 76722-388 76722-390 77090-138 77304-112
Methyl m-anisate 97%
Methyl m-anisate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
Boiling Pt: 252.0±0.0 °C (760 mm Hg)
Flash Pt: 93.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00017193
CAS Number: 5368-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR