Methyl (methoxymethylene)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-methoxyacrylate

A127805-100G A127805-10G A127805-500G A127805-25G A127805-5G
76718-820EA 33.95 USD
76718-820 76718-822 76718-824 77313-276 77313-278
Methyl (methoxymethylene)acetate 98%
Methyl (methoxymethylene)acetate

Precaución: for Research Use Only

Formula: C₅H₈O₃
MW: 116.1152 g/mol
Boiling Pt: 155.3 °C (760 mm Hg)
Melting Pt: 3…4 °C
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00128206
CAS Number: 34846-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR