Methyl 3-iodopyrazine-2-carboxylate 98%

Supplier: Ambeed
Warning

A373721-1G A373721-250MG
103826-540EA 70 USD
103826-540 103837-558 76840-826 76876-566
Methyl 3-iodopyrazine-2-carboxylate 98%
Methyl 3-iodopyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅IN₂O₂
MW: 264.021 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195309
CAS Number: 173290-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR