Methyl-3-iodo-2-methylbenzoate 97%

Supplier: Ambeed
Warning

A215070-100MG A215070-250MG A215070-5G A215070-10G
103818-452EA 10 USD
103818-452 103849-156 76873-786 76873-788 77087-680 77593-398
Methyl-3-iodo-2-methylbenzoate 97%
Methyl 3-iodo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉IO₂
Boiling Pt: 152 °C (14 mm Hg)
Melting Pt: 23…25 °C
Flash Pt: 130.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12923250
CAS Number: 52570-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR