Methyl-3-hydroxypropanoate 97%

Supplier: Ambeed
Warning

A365233-100G A365233-10G A365233-500G A365233-1G A365233-5G
103826-378EA 7 USD
103826-378 103837-398 76724-670 76724-672 76724-674 76840-636 77306-764
Methyl-3-hydroxypropanoate 97%
Methyl-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₄H₈O₃
Boiling Pt: 180.5 °C (760 mm Hg)
Flash Pt: 72.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00272293
CAS Number: 6149-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR