Methyl-3-hydroxycyclobutanecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-hydroxycyclobutanecarboxylate, Methyl 3-hydroxycyclobutane-1-carboxylate

A244733-100G A244733-10G A244733-500G A244733-1G A244733-25G A244733-5G
103811-600EA 7 USD
103811-600 103851-434 76815-498 76815-500 76815-502 77090-644 77090-646 77090-648
Methyl-3-hydroxycyclobutanecarboxylate 95%
Methyl-3-hydroxycyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
Boiling Pt: 190 °C (760 mm Hg)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00512755
CAS Number: 4934-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR