Methyl-3-hydroxybenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester

A196921-1000G A196921-100G A196921-10G A196921-500G A196921-25G A196921-5G
103847-492EA 9 USD
103847-492 77030-014 77030-016 77030-018 77030-020 77085-466 77302-542
Methyl-3-hydroxybenzoate 97%
Methyl-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₈O₃
Boiling Pt: 283.3 °C (760 mm Hg)
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002295
CAS Number: 19438-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR