Methyl-3-hydroxy-4-nitrobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-hydroxy-4-nitrobenzoate
A162224-100G
A162224-10G
A162224-500G
A162224-1G
A162224-25G
103805-590EA
5
USD
103805-590
103843-840
77235-890
77235-892
77235-894
77319-876
77319-878
Methyl-3-hydroxy-4-nitrobenzoate 97%
Methyl-3-hydroxy-4-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₈H₇NO₅ Flash Pt: 163.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016997 CAS Number: 713-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...