Methyl-3-hydroxypyrazine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Hydroxy-2-pyrazinecarboxylate

A150523-100G A150523-10G A150523-250MG A150523-1G A150523-25G A150523-5G
103804-176EA 10 USD
103804-176 103842-312 76719-702 76719-704 76719-706 77317-828 77317-830 77317-832
Methyl-3-hydroxypyrazine-2-carboxylate 98%
Methyl-3-hydroxypyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.1234 g/mol
Boiling Pt: 417.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03093338
CAS Number: 27825-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR