3H-Imidazo[4,5-c]pyridine-6-carboxylic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3H-imidazo[4,5-c]pyridine-6-carboxylate

A419027-250MG A419027-100MG
103827-366EA 127 USD
103827-366 103838-410 76877-102 77884-088
3H-Imidazo[4,5-c]pyridine-6-carboxylic acid methyl ester 95%
3H-Imidazo[4,5-c]pyridine-6-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656494
CAS Number: 82523-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR