Methyl-3-formylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-formylbenzoate

A242705-1G A242705-100G A242705-10G A242705-250MG A242705-25G A242705-5G
103811-486EA 7 USD
103811-486 103851-304 77090-488 77261-828 77261-830 77261-832 77304-252 77304-254
Methyl-3-formylbenzoate 98%
Methyl-3-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.1600 g/mol
Boiling Pt: 272.8 °C (760 mm Hg)
Melting Pt: 48…52 °C
Flash Pt: 118.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00189379
CAS Number: 52178-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR