Methyl 3-formyl-4-nitrobenzoate 97%

Supplier: Ambeed
Warning

A228914-100MG A228914-10G A228914-250MG A228914-5G A228914-1G A228914-25G
103819-428EA 13 USD
103819-428 76874-192 76874-194 76874-196 76874-198 77089-146 77089-148
Methyl 3-formyl-4-nitrobenzoate 97%
Methyl 3-formyl-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₇NO₅
MW: 209.16 g/mol
Melting Pt: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD05664271
CAS Number: 148625-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR