Methyl-3-fluoro-2-thiophenecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-fluorothiophene-2-carboxylate

A115752-10G A115752-250MG A115752-25G A115752-1G A115752-5G
103798-618EA 20 USD
103798-618 103824-478 77026-082 77026-084 77026-086 77297-392 77311-296
Methyl-3-fluoro-2-thiophenecarboxylate 95%
Methyl-3-fluoro-2-thiophenecarboxylate

Precaución: for Research Use Only

Formula: C₆H₅FO₂S
Boiling Pt: 216.791 °C (760 mm Hg)
Melting Pt: 51…53 °C
Flash Pt: 84.913 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16606491
CAS Number: 100421-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR