Methyl-3-fluoropicolinate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-pyridine-2-carboxylic acid methyl ester, Methyl 3-fluoropyridine-2-carboxylate

A196979-1G A196979-5G A196979-250MG
103816-882EA 17 USD
103816-882 103847-496 77085-472 77085-474 77112-016
Methyl-3-fluoropicolinate 98%
Methyl 3-fluoropicolinate

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.1300 g/mol
Boiling Pt: 228.8 °C (760 mm Hg)
Flash Pt: 92.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06659496
CAS Number: 869108-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR