Methyl-3-fluoro-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

A624010-25G A624010-100G A624010-250MG A624010-1G
103840-234EA 129 USD
103840-234 76844-402 76980-646 76980-648 77307-992
Methyl-3-fluoro-2-nitrobenzoate 98%
Methyl 3-fluoro-2-nitrobenzoate

Precaución: For research use only

Formula: C₈H₆FNO₄
Storage Temperature: Ambient
MDL Number: MFCD11520393
CAS Number: 1214353-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR