Methyl 3-chloro-5-methylpyrazine-2-carboxylate 95%

Supplier: Ambeed
Warning

A101677-250MG A101677-100MG
103796-088EA 339 USD
103796-088 76821-252 77884-922
Methyl 3-chloro-5-methylpyrazine-2-carboxylate 95%
Methyl 3-chloro-5-methylpyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
Boiling Pt: 265.6±35.0 °C (760 mm Hg)
Melting Pt: 84…85 °C
Flash Pt: 114.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697746
CAS Number: 859063-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR