Methyl-3-chloro-4-fluorobenzoate 98%

Supplier: Ambeed
Warning

A130002-100G A130002-1G A130002-25G A130002-5G
103801-364EA 6 USD
103801-364 76972-020 77313-786 77313-788 77313-790
Methyl-3-chloro-4-fluorobenzoate 98%
Methyl-3-chloro-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
Boiling Pt: 238 °C (760 mm Hg)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD06203675
CAS Number: 234082-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR