Methyl malonyl chloride 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 3-chloro-3-oxopropanoate
A385996-1G
A385996-25G
A385996-5G
A385996-10G
103826-790EA
11
USD
103826-790
103837-830
76841-178
76841-180
76841-182
77101-748
Methyl malonyl chloride 95%
Methyl malonyl chloride
Precaución: for Research Use Only
Formula:
C₄H₅ClO₃ MW: 136.53 g/mol Boiling Pt: 57…59 °C (12 mm Hg) Flash Pt: 80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013657 CAS Number: 37517-81-0 UN: 3265 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...