Methyl-3-bromothiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromothiophene-2-carboxylate

A255746-1G A255746-10G
103812-208EA 10 USD
103812-208 103852-068 77091-528 77685-727
Methyl-3-bromothiophene-2-carboxylate 98%
Methyl-3-bromothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅BrO₂S
MW: 221.07 g/mol
Boiling Pt: 251.7 °C (760 mm Hg)
Melting Pt: 50 °C
Flash Pt: 106 °C
Storage Temperature: Freezer
MDL Number: MFCD00173839
CAS Number: 26137-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR