3-Bromo-1H-pyrrole-2-carboxylic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Bromopyrrole-2-carboxylate

A756731-1G A756731-5G A756731-250MG A756731-25G
103840-874EA 77 USD
103840-874 76845-106 76845-108 77158-912 77158-914
3-Bromo-1H-pyrrole-2-carboxylic acid methyl ester 95%
3-Bromo-1H-pyrrole-2-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂
MW: 204.03 g/mol
Boiling Pt: 286.2±20.0 °C (760 mm Hg)
Flash Pt: 126.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD23106341
CAS Number: 941714-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR