Methyl 3-bromopropiolate 95%

Supplier: Ambeed
Danger

A180014-100MG A180014-250MG A180014-5G A180014-1G A180014-10G
103807-496EA 25 USD
103807-496 103845-850 76985-730 76985-732 77083-328 77301-658 77885-385
Methyl 3-bromopropiolate 95%
Methyl 3-bromopropiolate

Precaución: for Research Use Only

Formula: C₄H₃BrO₂
Boiling Pt: 150.2 °C (760 mm Hg)
Melting Pt: 20 °C
Flash Pt: 44.6 °C
Storage Temperature: Freezer
MDL Number: MFCD22989330
CAS Number: 23680-40-2
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR