Methyl-3-bromopropionate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-bromopropanoate
A123615-100G
A123615-10G
A123615-25G
A123615-500G
A123615-5G
A123615-1000G
103808-278EA
183
USD
103808-278
77094-594
77094-596
77094-598
77094-600
77094-602
77312-542
Methyl-3-bromopropionate 98%
Methyl-3-bromopropionate
Precaución: for Research Use Only
Formula:
C₄H₇BrO₂ Boiling Pt: 166.5 °C (760 mm Hg) Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000250 CAS Number: 3395-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...