Methyl-3-bromoisonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromoisonicotinate

A272551-1G A272551-25G A272551-5G A272551-100G A272551-10G
103813-104EA 5 USD
103813-104 103853-016 77092-796 77092-798 77092-800 77276-802 77276-804
Methyl-3-bromoisonicotinate 98%
Methyl 3-bromoisonicotinate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
Boiling Pt: 246.7 °C (760 mm Hg)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD03788226
CAS Number: 59786-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR