Methyl-3-bromoimidazo[1,2-a]pyridine-7-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate

A196044-100MG A196044-250MG
103816-806EA 76 USD
103816-806 103847-420 76721-280 76721-282
Methyl-3-bromoimidazo[1,2-a]pyridine-7-carboxylate 95%
Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566741
CAS Number: 342613-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR