Methyl 3-bromo-6-(trifluoromethyl)picolinate 98%

Supplier: Ambeed
Warning

A274794-100MG A274794-250MG A274794-1G A274794-5G
103813-224EA 135 USD
103813-224 103853-152 77032-004 77032-006 77305-180 77305-182
Methyl 3-bromo-6-(trifluoromethyl)picolinate 98%
Methyl 3-bromo-6-(trifluoromethyl)picolinate

Precaución: for Research Use Only

Formula: C₈H₅BrF₃NO₂
MW: 284.03 g/mol
Boiling Pt: 279.6±40.0 °C (760 mm Hg)
Flash Pt: 122.9 °C
Storage Temperature: Ambient
MDL Number: MFCD14698134
CAS Number: 1211538-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR