Methyl 3-bromo-5-fluoro-4-methylbenzoate 97%
Supplier: Ambeed
|
Warning
|
A248013-100MG
A248013-10G
A248013-250MG
A248013-1G
A248013-5G
103811-810EA
98
USD
103811-810
103851-646
77165-924
77165-926
77165-928
77304-436
77304-438
Methyl 3-bromo-5-fluoro-4-methylbenzoate 97%
Methyl 3-bromo-5-fluoro-4-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrFO₂ MW: 247.06 g/mol Boiling Pt: 272.9±40.0 °C (760 mm Hg) Flash Pt: 118.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD24107117 CAS Number: 1533932-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...