Methyl-3-bromo-5-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-5-(trifluoromethyl)benzoate

A418883-1G A418883-25G A418883-5G A418883-100G A418883-10G
103827-364EA 9 USD
103827-364 103838-408 76842-020 76842-022 76842-024 76979-230 76979-232
Methyl-3-bromo-5-(trifluoromethyl)benzoate 98%
Methyl 3-bromo-5-(trifluoromethyl)benzoate

Precaución: For research use only

Formula: C₉H₆BrF₃O₂
MW: 283.0420 g/mol
Boiling Pt: 248.9 °C (760 mm Hg)
Flash Pt: 104.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11847338
CAS Number: 187331-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR