Methyl 3-Bromo-5-(hydroxymethyl)benzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-5-(hydroxymethyl)benzoate

A254726-100MG A254726-10G A254726-250MG A254726-1G A254726-5G
103812-160EA 46 USD
103812-160 103852-012 76987-982 76987-984 76987-986 77091-458 77091-460
Methyl 3-Bromo-5-(hydroxymethyl)benzoate 95%
Methyl 3-Bromo-5-(hydroxymethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Boiling Pt: 354.2±32.0 °C (760 mm Hg)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD11040228
CAS Number: 307353-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR