Methyl 3-bromo-4-nitrobenzoate 97%

Supplier: Ambeed
Warning

A133609-250MG A133609-25G A133609-1G A133609-5G
103801-888EA 27 USD
103801-888 76823-124 76823-126 77314-560 77314-562
Methyl 3-bromo-4-nitrobenzoate 97%
Methyl 3-bromo-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
Storage Temperature: Ambient
MDL Number: MFCD11045599
CAS Number: 126759-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR