Methyl-3-bromo-4-iodobenzoate 98%
Supplier: Ambeed
|
Warning
|
A138636-100G
A138636-10G
A138636-250MG
A138636-1G
A138636-25G
A138636-5G
103802-644EA
7
USD
103802-644
77272-370
77272-372
77272-374
77299-004
77315-688
77315-690
Methyl-3-bromo-4-iodobenzoate 98%
Methyl 3-bromo-4-iodobenzoate
Precaución: for Research Use Only
Formula:
C₈H₆BrIO₂ Boiling Pt: 328.1 °C (760 mm Hg) Melting Pt: 56…60 °C Flash Pt: 152.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07778512 CAS Number: 249647-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...