Methyl-3-bromopicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-2-pyridinecarboxylate

A101417-100G A101417-10G A101417-25G A101417-500G A101417-1G A101417-5G
103795-966EA 15 USD
103795-966 103821-650 76780-498 76780-500 76780-502 76780-504 77282-728 77297-100
Methyl-3-bromopicolinate 98%
Methyl 3-bromopicolinate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
Boiling Pt: 267.4 °C (760 mm Hg)
Flash Pt: 115.533 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12025946
CAS Number: 53636-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR