Methyl-3-bromopyrazine-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A201381-10G
A201381-250MG
A201381-1G
A201381-5G
103817-288EA
25
USD
103817-288
103847-926
76785-350
76785-352
77085-998
77086-000
Methyl-3-bromopyrazine-2-carboxylate 95%
Methyl-3-bromopyrazine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O₂ Boiling Pt: 264.7 °C (760 mm Hg) Melting Pt: 44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08753935 CAS Number: 51171-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...