Methyl 3-bromo-2-methoxybenzoate 98%

Supplier: Ambeed
Warning

A216876-250MG A216876-1G
103818-584EA 59 USD
103818-584 103849-308 76826-062 77303-438
Methyl 3-bromo-2-methoxybenzoate 98%
Methyl 3-bromo-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042778
CAS Number: 260806-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR