Methyl 3-bromo-2-hydroxybenzoate 98%

Supplier: Ambeed
Warning

A181824-250MG A181824-5G A181824-1G A181824-25G
103807-658EA 15 USD
103807-658 103846-030 76985-780 77083-558 77301-750 77301-752
Methyl 3-bromo-2-hydroxybenzoate 98%
Methyl 3-bromo-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566798
CAS Number: 28165-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR