Methyl-3-bromo-2-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-2-fluorobenzoate

A120230-100G A120230-10G A120230-250MG A120230-25G A120230-1G A120230-5G
103799-902EA 12 USD
103799-902 76971-446 76971-448 76971-450 76971-452 77312-006 77312-008
Methyl-3-bromo-2-fluorobenzoate 98%
Methyl 3-bromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
Boiling Pt: 255.4 °C (760 mm Hg)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD09261255
CAS Number: 206551-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR