Methyl 3-bromo-2,4-dimethylbenzoate 98%

Supplier: Ambeed
Warning

A170393-100MG A170393-250MG A170393-5G
103806-484EA 78 USD
103806-484 76985-380 76985-382 77890-097
Methyl 3-bromo-2,4-dimethylbenzoate 98%
Methyl 3-bromo-2,4-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22570369
CAS Number: 151859-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR