Methyl-3-bromo-1H-indazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-1H-indazole-5-carboxylate

A157062-1G A157062-10G
103804-956EA 32 USD
103804-956 103843-158 77318-972 77592-924
Methyl-3-bromo-1H-indazole-5-carboxylate 97%
Methyl 3-bromo-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
Boiling Pt: 399.7±22.0 °C (760 mm Hg)
Flash Pt: 195.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11045401
CAS Number: 1086391-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR