Methyl 3-aminothieno[3,2-c]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A204475-1G A204475-100MG A204475-250MG
103817-554EA 142 USD
103817-554 103848-218 77086-390 77274-966 77274-968
Methyl 3-aminothieno[3,2-c]pyridine-2-carboxylate 95%
Methyl 3-aminothieno[3,2-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MW: 208.24 g/mol
Boiling Pt: 393.8±37.0 °C (760 mm Hg)
Melting Pt: 225…227 °C
Flash Pt: 192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234760
CAS Number: 111042-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR