Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-aminothieno[2,3-b]pyridine-2-carboxylate

A553194-250MG A553194-1G
103828-700EA 34 USD
103828-700 103839-832 76726-470 76843-914
Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate 98%
Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MW: 208.24 g/mol
Melting Pt: 198…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00174679
CAS Number: 111042-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR