Methyl-3-aminopicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-aminopyridine-2-carboxylate, Methyl 3-aminopicolinate

A193380-1G A193380-5G A193380-100G A193380-10G A193380-250MG
103816-528EA 23 USD
103816-528 103847-142 77085-046 77085-048 77274-536 77274-538 77274-540
Methyl-3-aminopicolinate 98%
Methyl 3-aminopicolinate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
Boiling Pt: 317.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08695602
CAS Number: 36052-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR